Learn More
Thermo Scientific Chemicals Quartz disc, fused, 50.8mm (2.0in) dia x 3.18mm (0.13in) thick
Product Code. 15466455
Change view
Click to view available options
Diameter (Metric):
50.8 mm
76.2 mm
Thickness:
1.59 mm
3.18 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Thickness Unit Size
15466455 76.2 mm 3.18 mm Each
15436455 50.8 mm 1.59 mm Each
15446455 50.8 mm 3.18 mm Each
15456455 76.2 mm 1.59 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15466455 Supplier Thermo Scientific Chemicals Supplier No. 042301.KS
 Restricted Item
Expédition estimée: 11-12-2026
Add to basket
This item is not returnable. View return policy
Manufactured by fusing naturally occurring crystalline silica
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mineral
Description Ground and polished
Diameter (Metric) 76.2 mm
Thickness 3.18 mm
Specimen Type Quartz Disc