Learn More
Quickfit™ Borosilicate Glass Large Capacity Soxhlet Extractor
Product Code. 15507810
Change view
Click to view available options
Capacity (Metric):
600 mL
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15507810 600 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15507810 Supplier Quickfit Supplier No. EX5/75
  • ONLINE EXCLUSIVE PRICE
    1240.00€
    1003.82€ / 2 pièces
    Économisez 236.18€
    19% de réduction
Expédition estimée: 07-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
With flat flange joints to permit introduction of large samples
  • Conforms to BS 2071
  • Large capacity extractors
  • With flat flange joints to permit introduction of large samples
  • Used in conjuction with flat flange lid and retaining clip which should be ordered separately
Specifications
For Use With (Equipment) Flat Flange Lid and Retaining Clip
Product Type Soxhlet Extractor
Capacity (Metric) 600 mL
Material Pyrex Borosilicate Glass
Cone Size 34/35
Certifications/Compliance BS 2071
Length (Metric) 370 mm