Learn More
Scilabware Quickfit™ Multiple Glass Joint Adapter with Two Parallel Necks
Product Code. 12021603
Change view
Click to view available options
Cone Size:
14/23
19/26
24/29
29/32
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Standard Taper Joint No. Unit Size
12021603 24/29 19/26 Each
12001603 14/23 14/23 Each
12425306 29/32 29/32 Each
12011603 19/26 19/26 Each
12031603 24/29 24/29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 12021603 Supplier Scilabware Supplier No. MA1/3
  • ONLINE EXCLUSIVE PRICE
    97.30€
    78.98€ / 1 pièces
    Économisez 18.32€
    18% de réduction
Expédition estimée: 01-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
Connect two apparatus together leak-tight. The QuickFit™ multiple glass joint adapter with two parallel necks is the perfect choice.
  • With two parallel necks
Specifications
Cone Size 24/29
Material Borosilicate Glass
Product Type Multiple neck adapter
Standard Taper Joint No. 19/26