Learn More
Quickfit™ Sloping Splash Head Adapter
Product Code. 15507980
Change view
Click to view available options
Cone Size:
19/26 (Condenser), 19/26 (Flask)
19/26 (Condenser), 24/29 (Flask)
24/29 (Condenser), 24/29 (Flask)
29/32 (Condenser), 29/32 (Flask)
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cone Size Unit Size
15507980 24/29 (Condenser), 24/29 (Flask) Pack of 5
15567970 29/32 (Condenser), 29/32 (Flask) Pack of 5
15577970 19/26 (Condenser), 19/26 (Flask) Pack of 5
15587970 19/26 (Condenser), 24/29 (Flask) Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15507980 Supplier Quickfit Supplier No. SH7/43
  • ONLINE EXCLUSIVE PRICE
    695.00€
    478.53€ / 5 pièces
    Économisez 216.47€
    31% de réduction
Expédition estimée: 23-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Used in distillation assemblies.
  • Used in distillation assemblies to prevent raw liquid carrying over from flask to condenser
Specifications
Material Borosilicate Glass (3.3)
For Use With (Application) Distillation assemblies to prevent raw liquid 'carrying over' from flask to condenser
Cone Size 24/29 (Condenser), 24/29 (Flask)
Product Type Sloping Splash Head