Learn More
(R)-1,2,3,4-Tetrahydronaphthalen-1-ol, 98%, BLD Pharm™
Product Code. 30948637
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948637 100 mg 100mg
30940947 250 mg 250mg
30932260 1 g 1g
30946779 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30948637 Supplier BLD Pharm Supplier No. BD12007100mg
 Restricted Item
Expédition estimée: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 23357-45-1
Molecular Formula C10H12O
MDL Number MFCD00063005
Quantity 100 mg
SMILES O[C@@H]1CCCC2=C1C=CC=C2
Molecular Weight (g/mol) 148.20
Percent Purity 98%
Chemical Name or Material (R)-1,2,3,4-Tetrahydronaphthalen-1-ol