Learn More
(R)-(+)-beta-methylphenethylamine, TRC
Product Code. 30346061
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346061 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346061 Supplier Toronto Research Chemicals Supplier No. TRCB3999382.5G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (R)-(+)-beta-methylphenethylamine
CAS 28163-64-6
Molecular Formula C9 H13 N
Quantity 2.5 g
Synonym (βR)-β-Methylbenzeneethanamine, ((R)-2-Phenylpropan-1-yl)amine, (+)-2-Phenylpropylamine, (+)-β-Methylphenethylamine, (2R)-1-Amino-2-phenylpropane, (2R)-2-Phenylpropan-1-amine, (2R)-2-Phenylpropan-1-amine, (2R)-2-Phenylpropylamine, (R)-(+)--Methylphenethylamine, (R)-(+)-2-Phenylpropylamine, (R)-2-Phenyl-1-propanamine, (R)-2-Phenyl-1-propylamine, (R)-2-Phenylpropylamine, (R)-2-Phenylpropylamine, (R)-β-Methylphenethylamine
InChI Formula InChI=1S/C9H13N/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/t8-/m0/s1
SMILES C[C@@H](CN)c1ccccc1
IUPAC Name (2R)-2-phenylpropan-1-amine
Molecular Weight (g/mol) 135.21
Formula Weight 135.1048