Learn More
rac-Nicotine-1,2',3',4',5',6'-13C6, TRC
Product Code. 30469545
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469545 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469545 Supplier Toronto Research Chemicals Supplier No. TRCN4124261MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material rac-Nicotine-1,2',3',4',5',6'-13C6
CAS 1189715-49-8
Molecular Formula 13C6 C4 H14 N2
Quantity 1 mg
Synonym rac-Nicotine (2,2',3,4,5,6-13C6), DL-Nicotine (2,3,4,5,6,2'-13C6), (±)-3-(1-Methyl-2-pyrrolidinyl-2-13C)pyridine-2,3,4,5,6-13C5, 3-(1-Methyl-2-pyrrolidinyl)pyridine-13C6, (±)-Nicotine-13C6, (±)-3-(1-Methyl-2-pyrrolidinyl)pyridine-13C6, (R,S)-Nicotine-13C6, (±)-Nicotine-13C6, 1-Methyl-2-(3-pyridyl)pyrrolidine-13C6, DL-Nicotine-13C6
InChI Formula InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/i2+1,4+1,6+1,8+1,9+1,10+1
SMILES CN1CCC[13CH]1[13c]2[13cH][13cH][13cH]n[13cH]2
IUPAC Name 3-(1-methyl(2^{13}C)azolidin-2-yl)(2,3,4,5,6-^{13}C_{5})pyridine
Molecular Weight (g/mol) 168.1875
Formula Weight 168.1358