Learn More
OHAUS™ Ranger™ Count 4000
Product Code. 15503893 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
3.00 kg
6.00 kg
15.00 kg
30.00 kg
Model:
RC41M15
RC41M15-M
RC41M3
RC41M3-M
RC41M30-M
RC41M6-M
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Model Unit Size
15503893 3.00 kg RC41M3 Each
15503903 15.00 kg RC41M15-M Each
15513893 15.00 kg RC41M15 Each
15513903 30.00 kg RC41M30-M Each
15583893 3.00 kg RC41M3-M Each
15593893 6.00 kg RC41M6-M Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15503893 Supplier OHAUS™ Supplier No. 30231102
  • ONLINE EXCLUSIVE PRICE
    610.00€
    440.84€ / 1 pièces
    Économisez 169.16€
    27% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
OHAUS™ Ranger™ Count 4000 is ideal for tough industrial counting jobs that require durability and accuracy. This scale is equipped with weighing, parts counting, check counting, and accumulation functions
  • Sturdy metal housing and slip-resistant rubber feet provide the protection, stability and long product life needed for tough industrial weighing.
  • Accurate weighing and counting performance is assured with 1:1,500,000 internal counting resolution and auto-optimization software that reduces errors.
  • Easy-to-operate with SmarText™ software, a centrally located count display and dedicated Sample and Tare keys.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 210 Hours with Rechargeable Battery
Connection Type Easy access communication port including standard RS232 (included) and second RS232, USB or Ethernet (accessories sold separately)
Construction Rugged Cast Aluminum Housing, Stainless Steel Platform
Capacity 3.00 kg
Display Type 3 Window Backlit LCD, 3 Color Checkweighing LEDs
Readability 0.100000 g
Linearity 0.200000 g
Model RC41M3
Repeatability 0.200000 g
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Gram, Kilogram, Pound, Ounce, Pound:Ounce
Dimensions (L x W x H) 327 x 315 x 122 mm
Weight (Metric) 15.500 kg