Learn More
Ohaus™ Ranger™ Count 4000 Counting Scale
Product Code. 15523893 Shop All Ohaus Products
Change view
Click to view available options
Capacity:
6 kg
30 kg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15523893 30 kg Each
15523883 6 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15523893 Supplier Ohaus™ Supplier No. 30231105
  • ONLINE EXCLUSIVE PRICE
    848.39€
    575.00€ / 1 pièces
    Économisez 273.39€
    32% de réduction
View all promotions
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Suitable for tough, industrial counting jobs that require durability and accuracy at a great value.
  • Equipped with weighing, parts counting, check counting, and accumulation functions, and can produce results for a variety of industrial weighing, counting, and measurement needs
  • Full metal housing offers strength and protection and maximizes product life
  • Precise counting and weighing performance produce accurate results
  • User-friendly software
  • Easy to operate
  • Several connectivity options provide connection to multiple peripheral devices
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 210 Hours with Rechargeable Battery
Connection Type 2nd RS232 (Available as an Accessory)Ethernet (Available as an Accessory)RS232 (Included)USB (Available as an Accessory)
Construction Rugged Cast Aluminum Housing, Stainless Steel Platform
Capacity 30 kg
Display Type 3-window Backlight LCD + 3 LED
Readability 1.000000 g
Includes RS232, AC Power, Rechargeable Battery
Linearity 2.000000 g
Material Rugged Cast Aluminum Housing, Stainless Steel Platform
Model RC41M30
Repeatability 2.000000 g
Dimensions (L x W) 327.00 x 315.00 mm
Dimensions (L x W x H) 327.00 x 315.00 x 122.000 mm
Length (English) 12.90 in.
Length (Metric) 327.00 mm
Length (Metric) Pan 225 mm
Width (English) 12.40 in.
Width (Metric) 315.00 mm
Width (Metric) Pan 300 mm
Height (English) 4.80 in.
Height (Metric) 122.000 mm
Depth (English) 12.90 in.
Depth (Metric) 327.00 mm
Electrical Requirements 100/240 VAC 50/60 Hz internal universal power supply, internal rechargeable lead acid battery