Learn More
Thermo Scientific™ Orion™ 2230XP Silica Analyzer Reagents and Standards
Product Code. 15914267
Change view
Click to view available options
Type:
200 ppb Calibration Standard
Reagents and Standard
Unit Size:
0.50 L
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15914267 Reagents and Standard 1 set
15904267 200 ppb Calibration Standard 0.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15914267 Supplier Thermo Scientific™ Supplier No. 2230RE
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
The reagents and calibration standards you need to operate and maintain the Thermo Scientific™ Orion™ 2230XP Silica Analyzer.
Ensure you have the reagents and calibration standards you need to operate and maintain the Thermo Scientific™ Orion™ 2230XP Silica Analyzer. Reagents kit contains all three reagents required for operation, and calibration standards available in individual bottles. For use with the Orion 2230XP Silica Analyzer.
Warranty: Out of box
TRUSTED_SUSTAINABILITY
Specifications
Description Reagents 1,2 and 3
Temperature (Metric) Operating 5°C to 45°C
Type Reagents and Standard
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.