Learn More
Recombinant VHH Alpaca, anti-Human, anti-Rabbit, Unconjugated, Clone: CTK0101, Proteintech ChromoTek
Product Code. 17313503
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17313503 500 μg 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17313503 Supplier Chromotek Supplier No. srbGCys21500
  • ONLINE EXCLUSIVE PRICE
    682.65€
    624.00€ / 500µL
    Économisez 58.65€
    8% de réduction
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Alpaca Monoclonal Antibody
500 μg Nano-Secondary™ for human IgG and rabbit IgG: alpaca monoclonal Nanobody, Fc-specific VHH, for 2x Cys conjugation, clone [CTK0101].
Specifications
Antigen Recombinant VHH
Applications Immunofluorescence, Western Blot, Flow Cytometry
Classification Monoclonal
Clone CTK0101
Conjugate Unconjugated
Host Species Alpaca
Quantity 500 μg
Primary or Secondary Secondary
Target Species Human, Rabbit