Learn More
Eppendorf™ Rectangular Centrifuge Bucket
Product Code. 11893882 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
5702 Centrifuge rotor A-4-38
Eppendorf rotor A-4-81 (CFA-152-020K)
Swinging bucket rotor CFA-146-010T (A-4-44)
Quantity:
2/Pk.
4/Pk.
Unit Size:
Pack of 2
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Quantity Unit Size
11893882 5702 Centrifuge rotor A-4-38 4/Pk. Pack of 4
10308612 5702 Centrifuge rotor A-4-38 2/Pk. Pack of 2
10136511 Swinging bucket rotor CFA-146-010T (A-4-44) 4/Pk. Pack of 4
10477272 Eppendorf rotor A-4-81 (CFA-152-020K) 4/Pk. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11893882 Supplier Eppendorf™ Supplier No. 5702709000
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Use these buckets with Eppendorf™ rotors.
  • Rectangular Centrifuge Buckets
  • Compatible with Eppendorf rotors
  • Available in packs of two and four
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 5702 Centrifuge rotor A-4-38
Capacity 90 mL
Container Type Bucket
Quantity 4/Pk.