Learn More
WTW™ Regeneration Set for WTW™ Oxygen Sensors CellOx 325, TA 197 Oxi, ConOx
Product Code. 10245681
Change view
Click to view available options
For Use With (Equipment):
Sensor CellOx 325 ConOx, TA 197 Oxi and MPP 350
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10245681 Sensor CellOx 325 ConOx, TA 197 Oxi and MPP 350 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10245681 Supplier WTW™ Supplier No. 202706
En stock
Add to basket
Edge
Add to basket
Membrane-covered galvanic dissolved oxygen sensors to measure in water and waterous solutions.
  • Regeneration Set for WTW™ Oxygen Sensors CellOx 325, TA 197 Oxi, ConOx
  • Membrane-covered galvanic dissolved oxygen sensors to measure in water and waterous solutions - low-maintenance and no polarization time, required
  • For instant measurement
  • Easy air calibration with water vapor saturated air in the calibration vessel
  • User friendly and long operation time
TRUSTED_SUSTAINABILITY
Specifications
Description Regeneration set
Includes Electrolyte, cleaning solution, 3 interchangeable heads foil diaphragm and grinding
For Use With (Equipment) Sensor CellOx 325 ConOx, TA 197 Oxi and MPP 350