Learn More
Rephile™ RephiDuo P Pack 1
Product Code. 15469719 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With (Equipment):
Water Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15469719 Water Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15469719
  • ONLINE EXCLUSIVE PRICE
    426.00€
    346.41€ / 1 pièces
    Économisez 79.59€
    18% de réduction
 Restricted Item
Expédition estimée: 08-02-2027
Add to basket
Add to basket
This product is designed to remove the most of the particles, organic compounds and chlorine from tap water to protect RO membranes.
RephiDuo P Pack 1 for Elix™ and Rios™ systems. Comparable to PR0G00001.
Compatible with following Millipore™ systems:
  • Direct-Q 3 /5™ (older version)
  • Elix (UV) 3/5/10™
  • RiOs 3™ (older version)
  • RiOs 5/8/16™
Disclaimer: These products are manufactured by Rephile. Rephile are not affiliated with or endorsed by Merck™.
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) Water Purification System
Type Cartidge Kit