missing translation for 'onlineSavingsMsg'
Learn More

Honeywell BW™ Replacement Oxygen (O2) Sensor

Product Code. 15839572
Change view
Click to view available options
Components Measured:
Oxygen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured
15839572 Oxygen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15839572 Supplier Honeywell Supplier No. SRX2V
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Honeywell BW™ Replacement Oxygen (O2) Sensor for GasAlert Extreme, GasAlertMicroClip XL, GasAlertQuattro, GasAlertMaxXT II, GasAlertMicro 5 Series

Specifications

Components Measured Oxygen
For Use With (Equipment) GasAlert Extreme, GasAlertMicroClip XL, GasAlertQuattro, GasAlertMaxXT II, GasAlertMicro 5 Series
Includes Sensor
Length (Metric) 5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.