Learn More
Moldex™ Respirator Fit Test Kit
Product Code. 12302309
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12302309 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12302309 Supplier Moldex™ Supplier No. 10301
  • ONLINE EXCLUSIVE PRICE
    433.93€
    281.00€ / 1 pièces
    Économisez 152.93€
    35% de réduction
Expédition estimée: 02-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Includes
Hood (1), Nebulisers (2), 6 x 2.5mL Bitrex™ Sensitivity Solution ampoules, 6 x 2.5mL Bitrex™ Test Solution ampoules, Spare parts for nebulisers (2), Instruction CD (1)
Allows employers to manage their own fit testing requirements
  • Fit test kit to perform qualitative fit tests
  • Contains everything needed for multiple fit tests
  • Free instruction CD
  • Packaged in sturdy carry case
Specifications
Product Type Fit Test Kit
For Use With (Application) Protection against dust, mite, and fume
Quantity 1 Kit