Learn More
Testo™ Robust Waterproof Immersion/Penetration Probe
Product Code. 10204603
Change view
Click to view available options
Diameter (Metric) Probe:
5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Probe Unit Size
10204603 5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10204603 Supplier Testo™ Supplier No. 0609 1273
  • ONLINE EXCLUSIVE PRICE
    138.50€
    93.00€ / 1 pièces
    Économisez 45.50€
    32% de réduction
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Type Probe, Robust Immersion/Penetration
Certifications/Compliance 60751 Standard
Response Time 12 sec.
Diameter (Metric) Probe 5 mm