Learn More
Horst™ Rod Sensors
Product Code. 10448641
Change view
Click to view available options
Diameter (Metric):
1mm
3mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10448641 3mm Each
10407701 1mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10448641 Supplier Horst™ Supplier No. 70250
  • ONLINE EXCLUSIVE PRICE
    140.90€
    89.50€ / 1 pièces
    Économisez 51.40€
    36% de réduction
Expédition estimée: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Diameter (Metric) 3mm
Diameter (Metric) Inner 3 mm
Length (Metric) 300 mm
Material PVC
Length (Metric) Power Cord 300mm
Product Type Rod Sensor
Temperature (Metric) 500°C