Learn More
Brady™ Roll Markers
Product Code. 11755573 Shop All Brady Products
Change view
Click to view available options
Display Text:
Air
Eau
Eau incendie
Vapeur
Dimensions (L x W x H):
33 m x 100 mm x 150 mm
33 m x 100 mm x 60 mm
Height (Metric) Character:
8 mm
13 mm
Unit Size:
1 roll
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Text Dimensions (L x W x H) Height (Metric) Character Unit Size
11755573 Air 33 m x 100 mm x 150 mm 13 mm 1 roll
11745573 Air 33 m x 100 mm x 60 mm 8 mm 1 roll
11775573 Vapeur 33 m x 100 mm x 60 mm 8 mm 1 roll
11765573 Eau 33 m x 100 mm x 60 mm 8 mm 1 roll
11735573 Eau incendie 33 m x 100 mm x 60 mm 8 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11755573 Supplier Brady™ Supplier No. 226384
  • ONLINE EXCLUSIVE PRICE
    363.00€
    306.67€ / 1 rouleaux
    Économisez 56.33€
    15% de réduction
Expédition estimée: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
Certifications/Compliance European Directives 67/548 and 88/379
Adhesive Type Permanent Acrylic
Coating Polyester
Color White on Blue
Display Pictogram and Text
Finish Opaque
Format Roll
Material Polyester
Product Type Pipe Marker
Sterility Non-sterile
Display Text Air
For Use With (Application) Pipe and valve marking
Languages French
No. per Pack 220 Markers/Roll
Quantity 220 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 100 mm
Dimensions (L x W x H) 33 m x 100 mm x 150 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 3.94 in.
Width (Metric) 100 mm
Height (English) 5.91 in.
Height (English) Character 0.51 in.
Height (Metric) 150 mm
Height (Metric) Character 13 mm
Alternate Part Number PMFL-AIR-BL4-RLL090