Learn More
Rosavin, TRC
Product Code. 30011836
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011836 5 mg 5mg
30003823 100 mg 100mg
30008393 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011836 Supplier Toronto Research Chemicals Supplier No. TRCR6849505MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84954-92-7
Molecular Formula C20 H28 O10
Molecular Weight (g/mol) 428.43
Synonym Rosavin, Rosavidin, (2E)-3-phenyl-2-propenyl 6-O-alpha-L-arabinopyranosyl-beta-D-Glucopyranoside
IUPAC Name (2R,3R,4S,5S,6R)-2-[(E)-3-phenylprop-2-enoxy]-6-[[(2S,3R,4S,5S)-3,4,5-trihydroxyoxan-2-yl]oxymethyl]oxane-3,4,5-triol
SMILES C(O[C@H]1[C@H](O)[C@@H](O)[C@@H](O)CO1)[C@H]2O[C@@H](OC/C=C/C3=CC=CC=C3)[C@H](O)[C@@H](O)[C@@H]2O
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C20H28O10/c21-12-9-28-19(17(25)14(12)22)29-10-13-15(23)16(24)18(26)20(30-13)27-8-4-7-11-5-2-1-3-6-11/h1-7,12-26H,8-10H2/b7-4+/t12-,13+,14-,15+,16-,17+,18+,19-,20+/m0/s1
Formula Weight 428.1682
Chemical Name or Material Rosavin