Learn More
Roseoflavin, TRC
Product Code. 30408980
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408980 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408980 Supplier Toronto Research Chemicals Supplier No. TRCR68500010MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Roseoflavin
CAS 51093-55-1
Purity Grade Notes HPLC
Molecular Formula C19 H24 N4 O6
Quantity 10 mg
InChI Formula InChI=1S/C19H24N4O6/c1-9-4-10-5-11-17(20-19(29)21-18(11)28)23(13(10)6-12(9)22(2)3)7-14(25)16(27)15(26)8-24/h4-6,14-16,24-27H,7-8H2,1-3H3,(H,21,28,29)/t14-,15+,16-/m0/s1
SMILES O[C@H]([C@H](O)[C@H](O)CO)CN(C1=NC2=O)C(C(N=C1C(N2)=O)=C3)=CC(N(C)C)=C3C
IUPAC Name 8-(dimethylamino)-7-methyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]pyrimido[4,5-b]quinoline-2,4-dione
Molecular Weight (g/mol) 404.42
Formula Weight 404.1696
Percent Purity >95