Learn More
Hirschmann™ rotarus™ Flow 50 Peristaltic Pump
Product Code. 15623516
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15623516 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15623516 Supplier Hirschmann™ Supplier No. 9501342
  • ONLINE EXCLUSIVE PRICE
    4483.50€
    3054.00€ / 1 pièces
    Économisez 1429.50€
    31% de réduction
View all promotions
Expédition estimée: 12-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
A comfortable, precise delivery pump with control options for velocity and flow speed.
  • For continuous delivery in speed ranges from 0.2 to 100 revolutions per minute
  • Performance class with 50 watt ensures exact delivery
  • White powder coating housing
  • Protection class IP 54
  • RFID technology is used to detect the pump head
  • Automatic blockage detection and hose rupture monitoring
  • Control panel for the most important functions is integrated directly into the housing
  • Equipped with an external control unit with touchscreen
  • Rapid and tool-free changing of pump heads
  • Operation with 24 V DC
Specifications
Enclosure Rating IP54
Motor Speed 0.2 to 100 rpm
Standard Connections USB and RS232 interface