Learn More
Horst™ Rubber Insulating Mat
Click to view available options
Diameter (Metric) Inner:
13 mm
25 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inner | 13 mm |
| Dimensions (L x W) | 2 Lm |
| Dimensions (L x W x H) | 2 Lm |
| Product Type | Insulating Mat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction