Learn More
Rubitecan, TRC
Product Code. 30420703
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420703 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420703 Supplier Toronto Research Chemicals Supplier No. TRCR70150010MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 9-Nitro-20(S)-Camptothecin
CAS 91421-42-0
Molecular Formula C20 H15 N3 O6
Quantity 10 mg
InChI Formula InChI=1S/C20H15N3O6/c1-2-20(26)13-7-16-17-10(8-22(16)18(24)12(13)9-29-19(20)25)6-11-14(21-17)4-3-5-15(11)23(27)28/h3-7,26H,2,8-9H2,1H3/t20-/m0/s1
SMILES CC[C@@]1(O)C(=O)OCC2=C1C=C3N(Cc4cc5c(cccc5nc34)[N+](=O)[O-])C2=O
Molecular Weight (g/mol) 393.35
Formula Weight 393.0961