Learn More
(S)-3-Aminopiperidin-2-one, TRC
Product Code. 30133391
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133391 500 mg 500mg
30009913 250 mg 250mg
30003948 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133391 Supplier Toronto Research Chemicals Supplier No. TRCA633018500MG
 Restricted Item
Expédition estimée: 09-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34294-79-6
Molecular Formula C5 H10 N2 O
Synonym (3S)-3-Amino-2-piperidinone∣(3S)-3-Aminopiperidin-2-one∣L-Orinithine Lactam;
Specifications
Quantity 500 mg
Shipping Condition Room Temperature
Formula Weight 114.0793
Molecular Weight (g/mol) 114.15
Chemical Name or Material (S)-3-Aminopiperidin-2-one