Learn More
(S,S)-Palonosetron Hydrochloride, TRC
Product Code. 30424168
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424168 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424168 Supplier Toronto Research Chemicals Supplier No. TRCP1658001G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Palonosetron Hydrochloride
CAS 135729-62-3
Molecular Formula C19 H24 N2 O . Cl H
Quantity 1 g
Synonym 1H-Benz[de]isoquinolin-1-one','2-[(3S)-1-azabicyclo[2.2.2]oct-3-yl]-2,3,3a,4,5,6-hexahydro-','hydrochloride (1:1)','(3aS)-, 1H-Benz[de]isoquinolin-1-one','2-(1-azabicyclo[2.2.2]oct-3-yl)-2,3,3a,4,5,6-hexahydro-','monohydrochloride','[S-(R*,R*)]-, 1H-Benz[de]isoquinolin-1-one','2-(3S)-1-azabicyclo[2.2.2]oct-3-yl-2,3,3a,4,5,6-hexahydro-','monohydrochloride','(3aS)- (9CI), Aloxi, Onicit, Palonosetron hydrochloride, RS 25259-197
InChI Formula InChI=1S/C19H24N2O.ClH/c22-19-16-6-2-4-14-3-1-5-15(18(14)16)11-21(19)17-12-20-9-7-13(17)8-10-20;/h2,4,6,13,15,17H,1,3,5,7-12H2;1H/t15-,17-;/m1./s1
SMILES Cl.O=C1N(C[C@H]2CCCc3cccc1c23)[C@@H]4CN5CCC4CC5
IUPAC Name (3aS)-2-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3a,4,5,6-tetrahydro-3H-benzo[de]isoquinolin-1-one;hydrochloride
Molecular Weight (g/mol) 332.87
Formula Weight 332.1655