missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Salmeterol xinafoate

Product Code. 15703467 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 15703467

Brand: Thermo Scientific Chemicals 462175000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94749-08-3
Molecular Formula C36H45NO7
Molecular Weight (g/mol) 603.76
MDL Number MFCD00897708
InChI Key XTZNCVSCVHTPAI-UHFFFAOYNA-N
Synonym salmeterol xinafoate, salmetedur, arial, serevent diskus, asmerole, betamican, dilamax, inaspir, ultrabeta, beglan
PubChem CID 56801
IUPAC Name 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol;1-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC=CC2=C1O.OCC1=CC(=CC=C1O)C(O)CNCCCCCCOCCCCC1=CC=CC=C1

Specifications

Melting Point 126.0°C to 128.0°C
Quantity 500 mg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmso, methanol
Formula Weight 603.75
Chemical Name or Material Salmeterol xinafoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.