Learn More
Velp Scientifica™ Sample Racks for Digestion Units
Product Code. 15379614 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
DK 42/26, DKl 42/26 Digesters
DK20 and DKL 20 Digesters
DK20, DKL20
DKL 20
DKL12
DKL12, 400 ml test tubes
DKL8
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15379614 DKL 20 Each
11795654 DK20 and DKL 20 Digesters Each
15309614 DK20, DKL20 Each
11775654 DKL12 Each
13487659 DKL12, 400 ml test tubes Each
11715664 DK 42/26, DKl 42/26 Digesters Each
11735654 DKL8 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15379614 Supplier Velp Scientifica™ Supplier No. A00000246
  • ONLINE EXCLUSIVE PRICE
    386.00€
    350.00€ / 1 pièces
    Économisez 36.00€
    9% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Facilitates nitrogen analysis and protein determination. Velp Scientifica™ Sample Racks for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
  • Available with or without heat shields
  • Available to fit a number of tube sizes and counts
Specifications
Product Type Sample Rack
For Use With (Equipment) DKL 20