Learn More
Thermo Scientific™ Sample and Skimmer Cones for iCAP™ MX and iCAP™ Q/Qnova Series ICP-MS Systems
Product Code. 17919660 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Sample Cone, Nickel
Skimmer Cone Insert 3.5 (HM)
Skimmer Cone, Ni, Insert
For Use With (Equipment):
iCAP MX and Q/Qnova ICP-MS
iCAP Q/Qnova Series ICP-MS
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes For Use With (Equipment) Unit Size
17919660 Sample Cone, Nickel iCAP MX and Q/Qnova ICP-MS Each
15457904 Skimmer Cone, Ni, Insert iCAP Q/Qnova Series ICP-MS Each
17807066 Skimmer Cone Insert 3.5 (HM) iCAP Q/Qnova Series ICP-MS -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17919660 Supplier Thermo Scientific™ Supplier No. 3600812
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
This item is not returnable. View return policy
Thermo Scientific™ sample and skimmer cones for Thermo Scientific™ iCAP™ MX and iCAP™ Q/Qnova series ICP-MS systems are available to suit your analysis requirements and designed for durability, performance and ease of use.
Assure robust, uncomplicated trace elemental analysis for your laboratory with the Thermo Scientific iCAP MX and iCAP Q/Qnova series ICP-MS systems. Our easily changeable Ni and Pt sample and skimmer cones are available to suit your analysis requirements and designed for durability, performance and ease of use.
Select high-quality consumables for consistent performance. These high-quality, field-proven cones are designed for reliable high-sensitivity analysis down to detection-limit level by ICP-MS. Available in either solid Ni construction or Pt tipped for even greater durability and specific analysis requirements.
  • Sample cones made from nickel and platinum
  • Skimmer cones made from nickel and platinum
  • Platinum, boron free skimmer cone (for semiconductor-related applications)
  • Skimmer cone inserts with different channel length to tailor instrument characteristics for each application
Specifications
Type Sample Cone
For Use With (Equipment) iCAP MX and Q/Qnova ICP-MS
Includes Sample Cone, Nickel