Learn More
Thermo Scientific™ Savant™ SpeedVac™ Refrigerated Vapor Trap Series
Product Code. 15996161 Shop All Thermo Scientific Products
Change view
Click to view available options
Electrical Requirements:
230 V 50 Hz
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Electrical Requirements Unit Size
15996161 230 V 50 Hz Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15996161 Supplier Thermo Scientific™ Supplier No. RVT450230
  • ONLINE EXCLUSIVE PRICE
    3840.00€
    3218.36€ / 1 pièces
    Économisez 621.64€
    16% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Condense and trap a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps.
Condenses and removes a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps (RVT). The RVT safely recovers the solvent and reduces emissions of hazardous chemicals in the environment.
Solvents are removed from samples during vacuum evaporation with a SpeedVac™ Concentrator which are then collected in a CFC-free Refrigerated Vapor Trap (RVT).   Solvent vapors pass into a Glass Condensation Flask and condense on its walls. This prevents the vapors from reaching and potentially damaging the oil-sealed vacuum pump. 
  • Rapid cooling time minimizes waiting period
  • Easy to remove glass flask for quick solvent changes
  • Rapidly cools to operating temperature
  • Easy to use glass condensation flask allows for the collection of liquid into a self-contained jar for easy draining
  • Sealed refrigeration system eliminates consumable coolants such as dry ice and acetone
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance UL Listed to US and Canadian requirements. CE marked
Description Refrigerated Vapor Cold Trap RVT450
Type Refrigerated Vapor Trap
Electrical Requirements 230 V 50 Hz