Learn More
SI Analytics™ ScienceLine Glass Single Reference Electrode
Product Code. 10315572 Shop All SI Analytics Products
Change view
Click to view available options
Reference Electrolyte:
Ag/AgCl
Calomel
Hg/Hg2SO4
Diameter (Metric) Body:
10 mm
12 mm
Length (Metric):
103 mm
120 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Reference Electrolyte Diameter (Metric) Body Length (Metric) Unit Size
10315572 Ag/AgCl 12 mm 120 mm Each
10213462 Calomel 10 mm 103 mm Each
10725013 Hg/Hg2SO4 10 mm 103 mm Each
10542113 Calomel 12 mm 120 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 10315572 Supplier SI Analytics™ Supplier No. 1070044
  • ONLINE EXCLUSIVE PRICE
    201.00€
    165.00€ / 1 pièces
    Économisez 36.00€
    17% de réduction
View all promotions
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Especially useful for the separation of combination electrodes into separate measuring and reference electrodes. SI Analytics™ ScienceLine Glass Single Reference Electrode are for special applications and make complex applications even more reliable.
  • The combination of pure measuring electrodes for i.e. ion-selective measurements or special constructions with one reference electrode
  • Redox titrations with a constant pH value with glass electrodes as reference and redox single sensors as measuring electrode
  • Application of double-electrolyte electrodes for an optimal adaptation of reference electrolyte to the measuring media.
TRUSTED_SUSTAINABILITY
Specifications
Description Ceramic Diaphragm
Material Glass
pH Range 0 to 14
Type Reference Electrode
Reference Electrolyte Ag/AgCl
Temperature (Metric) Range -5°C to +100°C
Length (Metric) 120 mm
Diameter (Metric) Body 12 mm