Learn More
3M™ Self Adjusting Breathing Tube
Product Code. 15779647 Shop All 3M Products
Change view
Click to view available options
Length (Metric):
52.5 to 85 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
15779647 52.5 to 85 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15779647 Supplier 3M™ Supplier No. 7000028067
  • ONLINE EXCLUSIVE PRICE
    66.00€
    52.42€ / 1 pièces
    Économisez 13.58€
    20% de réduction
Expédition estimée: 30-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
3M™ Self Adjusting Breathing Tube for 3M™ Adflo™ Powered Air Respirator and 3M™ Versaflo™ Supplied Air Regulator V-500E.
  • The new welding helmets and faceshields from 3M feature the QRS type of breathing tube (Quick Release Swivel), which enables one-handed connection and release
  • The protection class of Speedglas welding helmet 9100FX Air is reduced to TH2 when the adapter is used
  • You can extend the service life of the breathing tube by using a tube cover or a heavy-duty rubber breathing tube
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
Color Black
Product Type Breathing Tube
Length (Metric) 52.5 to 85 cm