Learn More
Brady™ Self-laminating Polyester Labels for M710
Product Code. 17837984 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
15 x 40 mm
22.86 x 22.86 mm
25.4 x 25.4 mm
38.1 x 15.24 mm
41.28 x 15.24 mm
44.45 x 25.4 mm
45.72 x 9.53 mm
48.26 x 15.24 mm
66.68 x 15.24 mm
66.68 x 25.4 mm
Color:
White
White, Transparent
Shape:
Rectangle
Rectangle with Circle
Square
Unit Size:
Pack of 100
Pack of 150
Pack of 250
Pack of 500
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Color Shape Unit Size
17837984 15 x 40 mm White, Transparent Rectangle with Circle Pack of 250
17867854 38.1 x 15.24 mm White, Transparent Rectangle Pack of 500
17808024 41.28 x 15.24 mm White, Transparent Rectangle Pack of 500
17827884 44.45 x 25.4 mm White, Transparent Rectangle Pack of 250
17888014 45.72 x 9.53 mm White, Transparent Rectangle Pack of 250
17867934 48.26 x 15.24 mm White, Transparent Rectangle Pack of 500
17898014 66.68 x 15.24 mm White, Transparent Rectangle Pack of 100
17817974 66.68 x 25.4 mm White, Transparent Rectangle Pack of 100
17848064 41.28 x 15.24 mm White, Transparent Rectangle with Circle Pack of 150
17808094 22.86 x 22.86 mm White Square Pack of 250
17808034 25.4 x 25.4 mm White, Transparent Square Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 17837984 Supplier Brady™ Supplier No. 311974
  • ONLINE EXCLUSIVE PRICE
    107.00€
    103.16€ / 250 pièces
    Économisez 3.84€
    3% de réduction
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Printer Labels for use with Brady™ Printer M710.
  • Self-laminating polyester has a permanent acrylic adhesive and is thermal transfer printable
  • Good print smudge resistance, solvent resistance and high/low temperature performance
Specifications
Certifications/Compliance ASTM D 1000
Adhesive Type Permanent Acrylic
Coating None
Color White, Transparent
Finish Matte
Format Label
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Self-laminating Tag
Sterility Non-sterile
For Use With (Application) Laboratory Labeling
For Use With (Equipment) M710, BMP71
Labels per Row 1 Label
Min. Operating Temperature (Metric) -196°C
Quantity 250 Labels
Shape Rectangle with Circle
Temperature Range (English) -321°F to +230°F
Temperature Range (Metric) -196°C to +110°C
Thickness (English) 0.0019 in.
Thickness (Metric) 0.048 mm
Dimensions (L x W) 15 x 40 mm
Length (English) 0.59 in.
Width (English) 1.57 in.
Width (Metric) 40 mm
Width (English) Printable Area 40 mm
Width (Metric) Printable Area 29.97 mm
Height (English) Printable Area 0.59 in.
Height (Metric) Printable Area 15 mm
Diameter (English) 0.375 in.
Diameter (Metric) 9.53 mm
Alternate Part Number M7-101-461