Learn More
Trajan™ GC Inlet Liner Accessories
Product Code. 11942637
Change view
Click to view available options
O-Ring, Viton; For All Liners Except SG092235
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11942637 O-Ring, Viton; For All Liners Except SG092235
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11942637 Supplier Trajan™ Supplier No. 0726532
Expédition estimée: 03-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Used as sealing rings for Agilent GC.
  • Suitable for Agilent GC
  • Temperature stable up to 300°C
Specifications
For Use With (Application) Viton O-Ring
For Use With (Equipment) Agilent, Thermo trace 1300 Bruker (Varian) Except SG092235
Material Viton
Quantity 10/Pk.
Product Type Viton O-ring for Agilent GC's