Learn More
Trajan™ Replacement Plungers for eVol and eVol NMR Syringes
Product Code. 12997596
Change view
Click to view available options
For Use With (Equipment):
eVol NMR Syringe (5μL)
eVol NMR Syringe (50μL)
eVol NMR Syringe (500μL)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12997596 eVol NMR Syringe (500μL) Each
12967596 eVol NMR Syringe (5μL) Each
12977596 eVol NMR Syringe (50μL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12997596 Supplier Trajan™ Supplier No. 2910384
Expédition estimée: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
SGE syringe replacement plungers for gas tight syringes
  • Replacement plungers for gas tight syringes
  • Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Product Type Replacement Plunger
For Use With (Equipment) eVol NMR Syringe (500μL)