Learn More
SGE™ Series II Standard Fixed Needle Syringes
Product Code. 10229010
Change view
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Needle Gauge:
25 G
26 G
Includes:
Plunger protection
Standard plunger
Quantity:
10/Pk.
6 Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 6
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Needle Gauge Includes Quantity Unit Size
10229010 50 μL 25 G Standard plunger Ea. Each
10518502 5 μL 26 G Plunger protection Ea. Each
12937536 10 μL 26 G Plunger protection 10/Pk. Pack of 10
10607171 10 μL 26 G Plunger protection Ea. Each
10127070 10 μL 26 G Plunger protection 6 Pk. Pack of 6
11941134 25 μL 25 G Standard plunger Ea. Each
10625471 100 μL 25 G Standard plunger Ea. Each
10559072 250 μL 25 G Standard plunger Ea. Each
10771422 500 μL 25 G Standard plunger Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10229010 Supplier SGE™ Supplier No. 004000
  • ONLINE EXCLUSIVE PRICE
    86.45€
    56.96€ / 1 pièces
    Économisez 29.49€
    34% de réduction
Expédition estimée: 15-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Standard syringes with anti-roll flange and needles that are aligned with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Stainless-steel plungers are hand fitted, noninterchangeable, nonreplaceable
  • Syringes with 5 and 10μL capacities have barrel O.D. of 6.5mm, scale length of 54.1mm and include a 50mm long 26s gauge needle
  • 25 to 500μL capacity syringes have 8mm O.D. barrel, 60mm scale length and 50mm long, 25 gauge needle Fixed-needle models have needle glued into barrel with solvent-resistant epoxy glue Removable-needle models feature a PTFE seal, a spring and nut for leak-resistant operation
Specifications
Accuracy ±1%
Material Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Needle Gauge 25 G
Diameter (English) Outer Needle 0.01 in.
Diameter (English) Outer 0.31 in.
Diameter (Metric) Outer 8 mm
Diameter (Metric) Outer Needle 0.5 mm
Includes Standard plunger
Length (English) Needle 1.96 in.
Product Type Microvolume Syringe
Graduations Major - 5.0 μL, Minor - 1.0 μL
Model 50F
Quantity Ea.
Volume (Metric) 50 μL
Product Line SGE
Length (Metric) Needle 50 mm
Needle Point Style Fixed Bevel
Connection Type Fixed
Graduated Yes