Learn More
SGI-1027, TRC
Product Code. 30420621
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420621 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420621 Supplier Toronto Research Chemicals Supplier No. TRCS2823755MG
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-[4-[(2-amino-6-methylpyrimidin-4-yl)amino]phenyl]-4-(quinolin-4-ylamino)benzamide
CAS 1020149-73-8
Molecular Formula C27 H23 N7 O
Quantity 5 mg
Synonym N-[4-[(2-Amino-6-methyl-4-pyrimidinyl)amino]phenyl]-4-(4-quinolinylamino)-benzamide
InChI Formula InChI=1S/C27H23N7O/c1-17-16-25(34-27(28)30-17)32-20-10-12-21(13-11-20)33-26(35)18-6-8-19(9-7-18)31-24-14-15-29-23-5-3-2-4-22(23)24/h2-16H,1H3,(H,29,31)(H,33,35)(H3,28,30,32,34)
SMILES Cc1cc(Nc2ccc(NC(=O)c3ccc(Nc4ccnc5ccccc45)cc3)cc2)nc(N)n1
IUPAC Name N-[4-[(2-amino-6-methylpyrimidin-4-yl)amino]phenyl]-4-(quinolin-4-ylamino)benzamide
Molecular Weight (g/mol) 461.518
Formula Weight 461.196