Learn More
Endecotts™ Sieve Calibration Ball
Product Code. 11994295
Change view
Click to view available options
Particle Size:
120 mesh
14 mesh
400 mesh
Unit Size:
Pack of 3
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Particle Size Unit Size
11994295 120 mesh Pack of 5
11924315 14 mesh Pack of 3
11934295 400 mesh Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11994295 Supplier Endecotts™ Supplier No. ZSICSA.125
  • ONLINE EXCLUSIVE PRICE
    281.00€
    270.94€ / 5 pièces
    Économisez 10.06€
    3% de réduction
 Restricted Item
Expédition estimée: 30-03-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Sieve balls, used to improve the sieving of cohesive material
    • Spheres are traceable to the United Kingdom National Physical Laboratory
    • Calibrate sieves to an accuracy of 1μm
    • Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture
    • Standards include a calibration chart
Specifications
Product Type Microspheres
Material Soda-lime glass
For Use With (Equipment) Sieve
Particle Size 120 mesh