Learn More
Sieve Shaker
Product Code. 13177913
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
13177913 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 13177913 Supplier Fritsch Supplier No. 03.8020.00
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
For accurate and reproducible sieving
Fritsch sieve shakers satisfy all requirements for accurate and reproducible sieving analysis and perform automatic sieve analysis with the control and evaluation software Autosieve. The Analysette 3 Pro guarantees absolutely reproducible sieving results due to automatic amplitude control, programme selection, can be integrated into any quality management system according to DIN EN ISO 9000, and has an RS232 interface.
Sieves, clamping lid, interposed sieve pans and sieve pans with and without outlet are also available for 100mm and 8inch sieves.Test sieves, as well as accessories for micro precision sieving and for use of the Analysette 3 as micro mill (for grinding small sample quantities), are available on request.
For quantitative particle size analysis of dry samples and suspensions of all kinds, by dry and wet sieving with woven test sieves.
Specifications
Voltage 230 V
Product Type Sieve Shaker
Frequency 50 Hz