Learn More
BRAND™ SILBERBRAND Automatic Burette with Lateral Safety Stopcock
Product Code. 10778431 Shop All Brand Products
Change view
Click to view available options
Includes:
2000 mL bottle
No bottle
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
10778431 2000 mL bottle Each
10034300 No bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10778431 Supplier BRAND™ Supplier No. 21911
Expédition estimée: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
SILBERBRAND Automatic Burette includes a lateral safety stopcock and black Schellbach stripe.
  • Calibrated to deliver (TD, Ex)
  • With needle-valve stopcock with PTFE spindle
  • Schellbach stripe
  • Automatic zeroing
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 10 mL
Graduations 0.02 mL
Includes 2000 mL bottle
Stopcock Lateral safety stopcock
Warranty 12 months