Learn More
Uvex Silv-Air Classic 2110 FFP1
Product Code. 30238146 Shop All Uvex Products
Change view
Click to view available options
Quantity:
15/Pk.
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30238146 15/Pk. Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30238146 Supplier Uvex Supplier No. 8762110
  • ONLINE EXCLUSIVE PRICE
    30.00€
    26.16€ / 15 pièces
    Économisez 3.84€
    12% de réduction
Expédition estimée: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Classic folding and cup-shaped respirators that fit all kinds of facial shape, with headband loops and integrated sealing lips in all protection classes.
  • Classic folding and cup-shaped respirators
  • Fit all kinds of facial shape
  • Headband loops and integrated sealing lips in all protection classes
  • Comfortable to wear for a longer period of time without becoming bothersome.
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type KN95
Material Polypropylene, Polyester
Product Type General Purpose Particulate Respirator
Size Medium
With Exhalation Valve Yes
Latex Free Yes
Quantity 15/Pk.