Learn More
CELLPATH SMALL SLIDE BOX + SLEEVE (FLATPACK)
Product Code. 30226312
Change view
Click to view available options
Color:
Brown
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30226312 Brown Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30226312 Supplier CELLPATH Supplier No. WES060008F
  • ONLINE EXCLUSIVE PRICE
    308.10€
    183.18€ / 25 pièces
    Économisez 124.92€
    40% de réduction
Expédition estimée: 07-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The robust Small Slide Box with Sleeve is suitable for archiving slides.
  • Key Benefits:
  • Made from recycled material
  • Bespoke boxes available at customer request
  • Cost efficient archive and storage
  • Dimension: 240 x 215 x 85 - holds 790 slides
Specifications
Type Slide Storage Box
Slide Capacity 790
Color Brown