Learn More
Gilac™ Smooth Wooden Handle
Product Code. 17988501
Change view
Click to view available options
Color:
Orange
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17988501 Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17988501 Supplier Gilac™ Supplier No. G020010
  • ONLINE EXCLUSIVE PRICE
    6.10€
    3.90€ / 1 pièces
    Économisez 2.20€
    36% de réduction
Expédition estimée: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Smooth Wooden Handle with 24 mm diameter is special for bakery and pastry.
  • Special for bakery and pastry.
Specifications
Product Type Broom Handle
Color Code Orange
Color Orange
Sterility Non-sterile
Autoclavable Not Autoclavable
Color-coded Yes
Material Wood
Diameter (Metric) 24 mm