Learn More
Sodium Cromoglicate EP Impurity A, Mikromol™
Product Code. 30050106
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050106 100 mg 100mg
30051187 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050106 Supplier Mikromol Supplier No. MM0039.03
 Restricted Item
Expédition estimée: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Sodium Cromoglicate EP Impurity A
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(2,6-Dihydroxyphenyl)ethanone (2',6'-Dihydroxyacetophenone)
CAS 699-83-2
Analyte or Component Names 1-(2,6-Dihydroxyphenyl)ethanone (2',6'-Dihydroxyacetophenone)
Shipping Condition Room Temperature
Molecular Formula C8 H8 O3
Certifications/Compliance ISO 9001
Synonym Sodium Cromoglicate Imp. A (EP), 1-(2,6-Dihydroxyphenyl)ethanone, 2',6'-Dihydroxyacetophenone, Sodium Cromoglicate Impurity A
InChI Formula InChI=1S/C8H8O3/c1-5(9)8-6(10)3-2-4-7(8)11/h2-4,10-11H,1H3
SMILES CC(=O)c1c(O)cccc1O
IUPAC Name 1-(2,6-dihydroxyphenyl)ethanone
Molecular Weight (g/mol) 152.15
Quantity 100 mg
Formula Weight 152.0473 g/mol