Learn More
Sodium hypophosphite monohydrate
Product Code. 11307707
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307707 250 g 250g
11317707 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307707 Supplier Thermo Scientific Alfa Aesar Supplier No. 014104.30
 Restricted Item
En stock
Add to basket
This item is not returnable. View return policy
CAS: 10039-56-2 | NaH{2}PO{2}^.H{2}O
Sodium hypophosphite monohydrate is used for electroless nickel plating agent and water treatment agent. It is also used in petroleum fields. Further, it is used as a food additive. In addition, it reduces nickel ions in solution to metallic nickel on metal substrates as well as on plastic substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 10039-56-2
Physical Form Solid
Quantity 250 g
Molecular Formula NaH{2}PO{2}^.H{2}O