Learn More
Sodium Methanethiosulfonate, TRC
Product Code. 30417736
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417736 10 g 10g
30417703 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30417736 Supplier Toronto Research Chemicals Supplier No. TRCS64500010G
 Restricted Item
Expédition estimée: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1950-85-2
Molecular Formula C H3 Na O2 S2
Molecular Weight (g/mol) 134.15
IUPAC Name methylsulfonylsulfanylsodium
SMILES CS(=O)(=O)S[Na]
Specifications
Quantity 10 g
InChI Formula InChI=1S/CH4O2S2.Na/c1-5(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1
Formula Weight 133.9472
Chemical Name or Material Sodium Methanethiosulfonate
Product Title
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.