Learn More
Sodium naphthalene-1-sulfonate, 85%, BLD Pharm™
Specifications
Specifications
| CAS | 130-14-3 |
| Molecular Formula | C10H7NaO3S |
| MDL Number | MFCD00064964 |
| Quantity | 1 kg |
| SMILES | O=S(C1=C2C=CC=CC2=CC=C1)([O-])=O.[Na+] |
| Molecular Weight (g/mol) | 230.22 |
| Percent Purity | 85% |
| Chemical Name or Material | Sodium naphthalene-1-sulfonate |