Learn More
behr Labor-Technik™ Soxhlet Extraction Equipment
Product Code. 10044200
Change view
Click to view available options
For Use With (Equipment):
Kompakt-system for extration and application-specific accessory kits
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10044200 Kompakt-system for extration and application-specific accessory kits Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10044200 Supplier behr Labor-Technik™ Supplier No. B00218114
  • ONLINE EXCLUSIVE PRICE
    6111.24€
    5011.50€ / 1 pièces
    Économisez 1099.74€
    17% de réduction
Expédition estimée: 22-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Order Info
ProductTypeSpecs
Specifications
Product Type Accessory Kits
For Use With (Application) the determination of free fat in foodstuffs
For Use With (Equipment) Kompakt-system for extration and application-specific accessory kits