Learn More
Socorex™ Spare Kit Bag, Socorex™ 187 Syringe
Product Code. 10633633
Change view
Click to view available options
For Use With (Equipment):
Socorex 187 Tube Feeding Syringe, 0.5 mL
Socorex 187 Tube Feeding Syringe, 10 mL
Socorex 187 Tube Feeding Syringe, 2 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10633633 Socorex 187 Tube Feeding Syringe, 10 mL Each
10580943 Socorex 187 Tube Feeding Syringe, 2 mL Each
10631903 Socorex 187 Tube Feeding Syringe, 0.5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10633633 Supplier Socorex™ Supplier No. 1.187.7210
Expédition estimée: 04-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Replace valve assembly parts for the Socorex™ 187 Tube Filling syringe if necessary. Socorex™ Spare Kit Bag, Socorex™ 187 Syringe includes all the spare parts needed for repair/replacement of your Socorex™ 187 Syringe valve.
  • Spare parts for Socorex™ 187 Tube Feeding Syringe models based on syringe volume.
  • Each bag includes the following:
    • 2/ea. springs.
    • 2/ea. balls.
    • 2/ea. valve washers.
    • 1/ea. barrel washer.
Specifications
Includes 2/ea. springs, 2/ea. balls, 2/ea. valve washers, one barrel washer.
Product Type Spare Kit Bag
Volume (Metric) 10 mL
For Use With (Equipment) Socorex 187 Tube Feeding Syringe, 10 mL