Learn More
3M™ Speedglas™ Head Protection 9100 FX
Product Code. 15889644 Shop All 3M Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15889644 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15889644 Supplier 3M™ Supplier No. 7100167240
  • ONLINE EXCLUSIVE PRICE
    136.00€
    108.08€ / 1 pièces
    Économisez 27.92€
    20% de réduction
Expédition estimée: 05-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
3M™ Speedglas™ Head Protection 9100 FX for use with Speedglas™ Welding Helmets 9100 FX and 9100 FX Air.
  • Made in flame-retardant textiles for protection against sparks and flames
  • Extended head coverage, for use with the complete helmet assembly to help protect the user from welding spark and spatter on the top portion of the head
  • As the owner of a Speedglas™ welding helmet, you have a variety of accessories available to help maximize your protection and comfort
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) Speedglas Welding Helmets 9100 FX and 9100 FX Air
Product Type Head Protection
Size Large
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.